Learn More
GA International CryoSTUCK TAPE – Cryogenic Tape for Frozen Surfaces
Supplier: GA International TFS25C150YE
Tape for the identification of previously frozen plastic containers, freshly removed from ultra-low temperature storage, even in the presence of frost and moisture. Ideal for long-term cryo-preservation of cryo vials, tubes, and freezer/cryo boxes in liquid or vapor phase liquid nitrogen (-196°C/-321°F), laboratory freezers (-80°C/-112°F), or transportation on dry ice. Provided in a continuous roll format, the cryo tape can be inscribed using permanent ink cryo markers. Available in an assortment of colors for easy identification and color coding.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.