Learn More
Cole-Parmer X-Ray Film Cassette, Aluminum, 5X7"
Supplier: Cole-Parmer 2170033
Cole-Parmer X-Ray Film Cassette, Aluminum, 5X7". Get the results you need at an affordable price-compare to name brand blue-sensitive, double-emulsion film for performance and value.Ideal for western blotting, DNA sequencing gels, chemiluminescent probes, gel shift analysis, and commonly used radioisotopes (32P, 125I)Double emulsion film gives you sharp contrast and exemplary resolution for DNA sequencingFilm is single-sided for exceptional clarity and low background.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.