Learn More
Thermo Scientific™ Xpect™ C. difficile Toxin A/B Test
Rapidly detect Clostridium difficile toxin A and B from human fecal samples with a simple, three-step procedure and results in 20 minutes.
Specifications
Content And Storage | 2°C to 30°C |
---|---|
Description | Xpect C. difficile Toxin A/B Test |
Includes | Test devices, conjugate reagent 1 (3.6mL), conjugate reagent 2 (3.0mL), specimen diluent, positive control (2.0mL), negative control (2.0mL), disposable transfer pipettes, dilution tubes, wooden applicator sticks |
Catalog Number | Mfr. No. | Description | Quantity | Content And Storage | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Quantity | Content And Storage | Price | Quantity & Availability | |||
R24640
|
Thermo Scientific™
R24640 |
Xpect C. difficile Toxin A/B Test | 40 Tests/Kit | 2°C to 30°C |
Pack of 40 for $2,371.00
|
N/A | |||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
R24650
|
Thermo Scientific™
R24650 |
Xpect C. difficile Toxin A/B Test | 20 Tests/Kit | 2°C to 30°C |
Pack of 20 for $1,619.00
|
N/A | |||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
Description
Rapidly detect Clostridium difficile toxin A and B from human fecal specimens using Thermo Scientific™ Xpect™ C. difficile Toxin A/B Test. Combine excellent clinical performance with a simple, three-step procedure that offers accurate results within 20 minutes with less than one minute hands-on time. When you need STAT testing or 24/7 coverage, choose Xpect.
Use this rapid, lateral flow immunoassay for the direct qualitative detection of Clostridium difficile toxin A and B. Enhance laboratory performance and efficiency with excellent clinical sensitivity and specificity and a simple walk-away procedure.Xpect C. difficile Toxin A/B uses antibody-coated colored particles to bind with target antigen in the diluted sample. The colored antibody/antigen complex moves along the test membrane. Immobilised antibodies specific to target antigen capture the color-labelled antigen complex at the test region, causing a clearly visible line to appear. This line will not appear in the absence of specific antigen. Immobilised, non-specific antibodies in the control region capture the color-labelled antibodies, whether antigen is present or not, resulting in a clearly visible control line.
- Clearly visible, easy-to-read results within 20 minutes
- Less than one minute hands-on time
- Integral control for added confidence
- Excellent sensitivity and specificity
Xpect Flu A&B
Xpect RSV
Xpect C. difficile Toxin A/B
Xpect Rotavirus
Xpect Giardia
Xpect Cryptosporidium
Xpect Giardia/Cryptosporidium
Xpect Legionella
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
2°C to 30°C | |
Xpect C. difficile Toxin A/B Test | |
Test devices, conjugate reagent 1 (3.6mL), conjugate reagent 2 (3.0mL), specimen diluent, positive control (2.0mL), negative control (2.0mL), disposable transfer pipettes, dilution tubes, wooden applicator sticks |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.