Learn More
Premier Lab Supply 45mm XRF Sample Cups – PN# SC-3345
Supplier: Premier Lab Supply SC3345
45mm Double-Open Ended XRF Sample Cup with Cup, Ring & Cap (PN# SC-3345) – Suitable for Universal – OD: 45mm; ID: 38mm; Height: 33mm; Volume: 20mL – 100 per bag