Learn More
Cole-Parmer Cole-Parmer Blue-Sensitive Autoradiography Film, 5x7", 100/pk
Supplier: Cole-Parmer 2170001
Ideal for western blotting, DNA sequencing gels, chemiluminescent probes, gel shift analysis, and commonly used radioisotopes (32P, 125I). Use for manual or automatic development.