Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Coast™ WX850R 700 Lumen Dual Color Rechargeable Focusing LED Headlamp

The WX850R is our most powerful rechargeable front loader, combining a Pure Beam Focusing Optic with a red beam option to maximize professional or recreational capability.

Supplier:  Coast™ 21554

Catalog No. 19-012-030

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.


Includes: Lithium battery pack, 3 x AAA batteries, USB charging cord, AC adapter, hardhat clips

Requires : Lithium-Ion Pack or 3 x AAA

Description

Description

The WX850R Headlamp is our most powerful rechargeable front loaded LED headlamp. The light features our Flex Charge Dual Power system. The Flex Charge feature allows for the rechargeable battery pack to be recharged inside or outside of the headlamp using a micro USB. The Dual Power allows for it to run on our custom rechargeable lithium battery pack or standard alkaline batteries. The first button on the headlamp gives you the ability to shine an ultra wide flood beam and then quickly twist the bezel to a long reaching spot beam with transition halo. The second button on the headlamp enables the user to switch quickly to a fixed red LED light to preserve night vision. It also includes a reflective safety strap for additional safety benefits.

With Coast’s commitment to quality, the WX850R Headlamp is impact resistant, weather resistant and its LED is virtually unbreakable. The WX850R Headlamp is backed by Coast’s lifetime warranty against defects in materials and workmanship. Tested and rated to ANSI/FL1 standards.

Certifications

ANSI/FL1 Standards

Compliance

ANSI/FL1 Standards

Warranty and Services

Lifetime

Specifications

Specifications

Rechargeable Headlamp
0.11 kg
21.2 cm
Plastic with Fabric Strap
0.24 lb.
Yellow, Gray
5.7 cm
14.2 cm
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit