Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Worldwide Glass Resource 13-425 Polypropylene Screw Thread Closures

Open-top closures for chromatography vials

$350.00 - $354.50

Specifications

Color Black
Material Polypropylene
Closure Size 13-425
For Use With (Equipment) Chromatography Vials
View More Specs

Products 2
Catalog Number Mfr. No. Pre-slit Price Quantity & Availability  
Catalog Number Mfr. No. Pre-slit Price Quantity & Availability  
14-812-128
View Documents
Worldwide Glass Resource
0016168
Yes
Case of 1000 for $354.50
Add to Cart
 
14-812-127
View Documents
Worldwide Glass Resource
0016150
No
Case of 1000 for $350.00
Add to Cart
 
Description

Description

  • 13-425 screw thread finish
  • Polypropylene construction

Specifications

Specifications

Black
Polypropylene
13-425
Chromatography Vials
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit