Learn More
Molecular Dimensions WIZARD CLASSIC 1 & 2 HT96
Supplier: Molecular Dimensions MD15W12B
Protein Crystallography. WIZARD CLASSIC 1&2 Screens HT-96 (96 x 1.7mL): designed to increase your probability of producing crystals during the coarse screening phase when crystallizing biological macromolecules (proteins, nucleic acids, peptides, and complexes).