Learn More
iQ Biosciences Wistar Rat PBMCs, 10M cells/vial
Supplier: iQ Biosciences IQBRPB202
High-quality, high-viability (> 80%) peripheral blood mononuclear cells (PBMCs) from the Wistar rat, a widely-used outbred rat strain. Used for a wide variety of immunology-based applications, including in-vitro immunoassays and ex-vivo applications. All orders come with an iQ Certificate of Analysis.