Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VAI WipeDown High Sorb Dry Wipes
WipeDown High Sorb Dry Wipes are high sorbancy laundered polyester wipes with laser cut sealed edges. Low particulate, low extractable wipes.
Supplier: VAI VEL812X12S3031
Description
- ISO 5 (Grade A/B, Former Class 100) Cleanliness
- Material is exceptionally clean and absorbent
- Resistance to abrasion and picking
Specifications
12 in. | |
12 in. | |
Polyester | |
20 Pk. | |
A/B |
30.5 cm | |
30.5 cm | |
ISO 5 | |
Sterile |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction