Learn More
Malvern Panalytical Window + Perlast Seals (Pair)
Supplier:  Malvern Panalytical MAZ2014
Encompass
Pair of anti-reflection coated windows and Perlast window seals for the Pair of anti-reflection coated windows and Perlast window seals for theMastersizer 3000 wet measurement cell (MAZ2010 or MAZ2020).Chemical Compatibility code A.
Catalog No. NC1213294