Learn More
Bel-Art™ SP Scienceware™ Wilmad™ Universal Solvent Jet NMR Tube Washer
GSA_VA
Cleans NMR tubes in small batches. Bel-Art SP Scienceware™ Universal Solvent Jet NMR Tube Washer can also be used for cleaning gas-tight sample tubes.
Manufacturer:  Bel-Art™ 660000086
Catalog No. 16-800-186
Add to cart
Description
Description
  • Loosen and retighten the fitting on the assembled washer head to extend the PTFE tubing into the sample tube.
  • Washer head affixes to a filter flask that includes a vacuum sidearm (sold separately)
  • When a vacuum is applied to the flask, the sample tube is pressed against the rubber gasket to form an air-tight seal that starts the solvent flow.
  • Sample tube can be air-dried by lifting the PTFE tubing out of the solvent reservoir
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Single Tube Washer
Fits tubing: 2.5 to 5.0mm O.D.
Washer Body, rubber gasket, PTFE tubing, and PTFE tubing adapter.
WG-7200-1