Learn More
Bel-Art™ SP Scienceware™ Pyrex™ Capillary Tubes
Manufactured from Pyrex™ and sealed at one end. Bel-Art SP Scienceware™ Wilmad™ Pyrex™ Capillary Tubes provide a low cost solution for small volume NMR measurement.
Supplier:  Bel-Art™ 660000073
 View more versions of this product
Catalog No. 16-800-173
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Description
Description
  • Made of ASTM Type 1 Class A glass
  • Tested at 400 MHz field
Specifications
Specifications
Pyrex™ Glass
100 mm
2.2 mm
4 in.
WG-1364-2.5A
Capillary Tube
2.5 mm
400MHz
10 Pack
Product Suggestions
Product Suggestions