Learn More
Bel-Art™ SP Scienceware™ Wilmad-LabGlass™ Replacement O-Ring for Spinner
Bel-Art™ SP Scienceware™ Wilmad-LabGlass™ Replacement O-Ring for Spinner is manufactured to precision standards that exceed spectrometer products in terms of dimension tolerance, reliability and life span.
Supplier: Bel-Art™ SP Scienceware™ 660000054
Description
- Low cost alternative to the full VT version for sample changers.
- Weight and susceptibility matched to originals.
- Can be mixed with originals in sample changer operation.
- Limited VT operation around ambient.
- Does not jam at top of upper barrel during insert operation.
- Tachometer strip now with foil protection for improved life span.
Specifications
Agilent/Varian Turbines | |
5 mm | |
Viton | |
TURBINE-ORING-RED |