Learn More
Bel-Art™ SP Scienceware™ Wilmad-LabGlass™ Open Cap for 4 to 5 mm Tubes
Bel-Art™ SP Scienceware™ Wilmad-LabGlass™ Open Cap for 4 to 5 mm Tubes are commonly used in sample degasification.
Supplier: Bel-Art™ SP Scienceware™ 660000053
Description
- Comes complete with open top screw cap
- Inert to most organic solvents and compounds but poor to strongly corrosive materials
- Remains reliable after multiple punctures
Specifications
Replacement Open Cap Only | |
Black | |
4 and 5 mm Screw Cap NMR Tubes |
Model: TR-OC-01 | |
12 pieces |