Learn More
Electronic Imaging Materials Inc 667 Microplate CryoLabel 2.25 inch x 0.25 inch, White, 1.5K labels per roll, 1 inch core
SDP
Supplier:  Electronic Imaging Materials Inc 667225025
Encompass_Preferred
2.25 inch x 0.25 inch Microplate CryoLabel will handle extremely cold storage temperatures found in genetics, virology, microbiology and related fields. Conformable and resistant to moisture, smudging, and tearing, plus it holds up to alcohol and other chemicals. Designed for thermal transfer and durable UV inkjet printing, allowing you to color-code labels. Scratch and smear resistant when printed with T68 premium resin ribbon.
Catalog No. NC1235361