Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Amerex™ Wheeled Fire Extinguishers: Regular Dry Chemical
Wheeled, dry chemical fire extinguisher
$8960.00 - $14570.00
Specifications
Length (English) Hose | 50 ft. |
---|---|
Length (Metric) Hose | 15 m |
Range | 30 to 40 ft. |
Reagent Type | Regular dry chemical |
Temperature Range (English) | -65° to +120°F |
Catalog Number | Mfr. No. | Capacity (English) | Dimensions (L x W x H) | Discharge Time | Product Type | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (English) | Dimensions (L x W x H) | Discharge Time | Product Type | Price | Quantity & Availability | ||
19-860-545
|
Amerex™
11403 |
150 lb. | 38 x 26 x 52 in. (96.5 x 66 x 132 cm) | 53 sec. | Semi-pneumatic Wheeled Fire Extinguisher |
Each for $10,840.00
|
|
19-860-551
|
Amerex™
15504 |
350 lb. | 39 x 34 x 57 in. (99 x 88 x 145 cm) | 70 sec. | Steel-wheeled Fire Extinguisher |
Each for $14,570.00
|
|
19-860-543
|
Amerex™
11616 |
150 lb. | 36 x 24 x 45 in. (91.4 x 61 x 114.3 cm) | 60 sec. | Semi-pneumatic Wheeled Fire Extinguisher |
Each for $8,960.00
|
|
19-860-548
|
Amerex™
471 |
150 lb. | 45 x 28 x 52 in. (114 x 71 x 132 cm) | 53 sec. | Steel-wheeled Fire Extinguisher | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
- Semi-pneumatic wheels
- Coast Guard approved
- Contains a siliconized sodium bicarbonate-based dry chemical with free flowing and non-caking additives
- Economical Class B and C protection with lower initial cost and recharging
- Chemical smothers fires in flammable liquids and pressurized gases and is electrically nonconductive
Specifications
50 ft. | |
30 to 40 ft. | |
-65° to +120°F |
15 m | |
Regular dry chemical | |
-54°C to +49°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title