Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytiva Whatman™ Puradisc™ 25mm Polypropylene Syringe Filters
Whatman™ Puradisc™ 25mm Polypropylene Syringe Filters may be used for biological sample and HPLC aqueous sample preparation.
$527.00 - $3530.00
Specifications
Sterility | Non-sterile |
---|---|
Filtration Area | 4.2 cm2 |
Volume (Metric) | Up to 100 mL |
Diameter (Metric) | 25 mm |
Catalog Number | Mfr. No. | Pore Size | No. per Pack | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Pore Size | No. per Pack | Price | Quantity & Availability | ||||
05-713-404A
|
Cytiva
67902504 |
0.45 μm | 100 |
Pack of 1000 for $3,530.00
|
|
||||
05-713-404
|
Cytiva
67882504 |
0.45 μm | 200 |
Pack of 200 for $1,341.00
|
|
||||
05-713-402
|
Cytiva
67862504 |
0.45 μm | 50 |
Pack of 50 for $527.00
|
|
||||
05-713-401
|
Cytiva
67862502 |
0.2 μm | 50 | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
05-713-403
|
Cytiva
67882502 |
0.2 μm | 200 | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
Description
Whatman™ Puradisc™ 25mm Polypropylene Syringe Filters may be used for biological sample and HPLC aqueous sample preparation.
- Diameter: 25mm
- Sample volume up to 100mL
- Low hold-up volumes for maximum sample recovery
- Optional glass microfiber
- Autoclave at 121ºC
Applications:
- HPLC aqueous sample preparation
- Biological sample preparation
- Buffer solutions
- Salt solutions
- Tissue culture media
- Irrigation solutions
- Sterile isolation
Specifications
Non-sterile | |
4.2 cm2 | |
Up to 100 mL | |
25 mm |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title