Learn More
Revvity Health Sciences Inc WGA PVT SPA 100 MG
Supplier: Revvity Health Sciences Inc RPNQ0252
SPA Scintillation beads are microspheres containing scintillant which emit light in the blue region of the visible spectrum. As a result, these beads are ideally suited to use with photomultiplier tube (PMT) counters such as the MicroBeta2 or TopCount. Two types of core SPA Scintillation bead are available - yttrium silicate (YSi) and Polyvinyltoluene (PVT). PVT beads are plastic, larger in size, and stay in suspension longer than the crystalline YSi beads. Scintillation proximity assay (SPA) is a homogeneous and versatile technology for the rapid and sensitive assay of a wide range of biological processes, including applications using enzyme and receptor targets, radioimmunoassays, and molecular interactions. When 3H, 14C, 33P, and 125I radioisotopes decay, they release ?-particles (or Auger electrons, in the case of 125I). The distance these particles travel through an aqueous solution is dependent on the energy of the particle. If a radioactive molecule is held in close enough prox
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.