Learn More
LI-COR Black Western Blot Incubation Boxes (small - 10 boxes)
Supplier: LI-COR 92997110
Black blot boxes are suitable for incubating near-infrared fluorescent and chemiluminescent Western blots or protein gels. Size: 7.2 cm x 4.9 cm x 3.0 cm