Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ FlexTech™ Y9290 Full NBR-Coated Cut-Resistant Gloves with Nitrile-Coated Palm

Catalog Number p-4703897
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

FlexTech™ Y9290 Full NBR-Coated Cut-Resistant Gloves with Nitrile-Coated Palm is a lightweight, cut resistant shell that is double coated with a layer of nitrile rubber (NBR) for water-resistance, and has an additional layer of nitrile coating on the palm and fingers for cut protection.

  • Lightweight, cut-resistant knit shell with double coating
  • Sandy nitrile coated palm and fingertips enhances grip
  • Full NBR coating provides an additional layer of protection against liquids
  • Gloves offer a balance of comfort, dexterity and cut resistance
  • Water resistant
ANSI Abrasion Rating ANSI 4
Certifications/Compliance ASTM F1790-97, ASTM D3389-10
Coated Area Palm, Fingertips
Construction Knit
Coating Material Nitrile
Color Gray
Cut Resistance Rating ANSI A4
Exterior Finish Fully Textured
Hand Specific
Material Nitrile Rubber
Product Type Cut-resistant Gloves
Size Small
EN388 Standard EN 388:4543
For Use With (Application) Construction, metal fabrication, assembly
Glove Cuff Color Red
Glove Cuff Style Knit
Model Y9290S
Puncture Rating ANSI/ISEA 105-16 Level 4
Series Flextech™ Y9290
Yarn Gauge 13
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit