Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Wearwell™ F.I.T. Functional Interlocking Tile
For use in mostly wet areas — surface slants toward holes to facilitate drainage
Supplier: Wearwell™ 540.58X12X12BKCS20
Description
- Angular “LockSafe” interlocking system provides a stronger connection, yet the tiles easily zip apart for re-positioning
- Ergonomic design and PVC formula reduce fatigue
- Easy ramp system: One ramp fits both male and female edges
- Tiles snap together easily and can be configured to fit your exact work spaces
- Save on replacement costs and installation time
- Available with gritworks™ that provides a slip-resistant work surface for use in oily areas
- Color: Black
Order Info
Beveled ramps and ramps with corners also available. Contact your Fisher Safety Customer Service Representative for ordering information.
Warranty and Services
One year
Specifications
0.63 in. | |
12 in. | |
Black | |
Anti-fatigue Flooring |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction