Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Wearwell™ 702 Corrugated Switchboard Matting
Supplier: Wearwell™ 702.14X4X75BK
Specifications
4 x 75 ft. | |
Black | |
0.25 in. | |
Insulates the standing worker from high voltage shock |
Non-conductive Mat | |
Corrugated Switchboard Matting | |
ANSI/ASTM D178-01, Type II, Class 2 | |
PVC |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction