Learn More
PATTERSON DENTAL COMPANY Patterson Wax Square Ropes – Size 11" Long x 3/16", White, 55/pk
Supplier: PATTERSON DENTAL COMPANY 0911503
Wax Square Ropes from Patterson are ideal for border molding impression trays, tacky but not greasy. Ropes are 3/16" square and 11" long.