Learn More
Wax Out™ Cleaning Solution, Fisher Chemical
Cleaning solvent for paraffin and other waxes used in histology. Contains kerosene and α-limonine
Description
Specifications
Liquid | |
175°C | |
Aluminum Can | |
4 L |
Wax Out™ Cleaning Solution | |
8008-20-6, 5989-27-5 | |
Yellow | |
MFCD00212819 |