Learn More
Labconco™ WaterPro™ BT Water Purification Systems



The WaterPro BT System is ideal for laboratories needing 1-10L/day, delivering both ultrapure Type I (18.2 megohm-cm) and reverse osmosis (RO) Type III water. Benchtop design is compact, lightweight, and quick to install. The WaterPro BT may be easily transported and shared among labs.
$7335.00 - $8552.00
Specifications
Certifications/Compliance | UL 61010-1; CAN/CSA C22.2 No. 61010.1; CE electrical safety and electromagnetic compatibility |
---|---|
Type | Water Purification System |
For Use With (Application) | Type I water: Instrumental trace element analysis, mixing standardized acid/base solutions, and media solutions Type III water: Non-critical laboratory applications such as reagent preparation and glassware rinsing |
Plug Type | 115 V, 15 A |
Width (English) | 11.4 in. |
Requires: WaterPro BT Filter Pack (9019200)
Catalog Number | Mfr. No. | Depth (Metric) | Height (Metric) | Includes | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Depth (Metric) | Height (Metric) | Includes | Price | Quantity & Availability | |||
S28405
![]() ![]() |
Labconco™
9015020/EMD |
42.1 cm | 53.8 cm | N/A |
Each for $7,335.00
|
|
S28403
![]() ![]() |
Labconco™
9015120/EMD |
42.1 cm | 53.8 cm | UV lamp |
Each for $8,552.00
|
|
Description
- Delivers reverse osmosis (RO)-purified Type III water at a rate up to 3L/hour (0.05L/min.)
- Delivers ultrapure Type I (18.2 megohm-cm) water at a typical rate of >0.5L/min. at an inlet water temperature of 25°C
- RO-purified water storage tank 6L capacity
- Thermoplastic housing with removable thermoplastic cover
- Powerful, efficient distribution pump with continuous/intermittent recirculation
- Color LCD for display of operating modes, operating parameters, maintenance or alarm messages, and tank fill level
- Visual alarms including filter cartridge change after set time has expired and volume of dispense water has been reached, resistivity falls below setpoint, incorrect filter installation, and water detected in tank while in Flush mode
- Keypad for accessing display settings, function settings, and calibrations
- 50 dB maximum noise level at a distance of 3.25 feet
- Feedwater inlet tubing, 7 ft. long with 1/2 in. OD male NPTM, BSPM or GAZ fitting and pre-filter screen
- RO reject tubing, 80 in. long
- One year warranty
- UV Lamp, 185 and 254 nanometers
- Remote Dispenser with 2m (6.5 ft.) tether
- White powder-coated steel dispenser stand
- UL 61010-1; CAN/CSA C22.2 No. 61010.1
- CE electrical safety and electromagnetic compatibility
Warranty and Services
1 year
Specifications
UL 61010-1; CAN/CSA C22.2 No. 61010.1; CE electrical safety and electromagnetic compatibility | |
Type I water: Instrumental trace element analysis, mixing standardized acid/base solutions, and media solutions Type III water: Non-critical laboratory applications such as reagent preparation and glassware rinsing | |
11.4 in. | |
21.2 in. | |
16.6 in. | |
115 V 60 Hz 2 A |
Water Purification System | |
115 V, 15 A | |
29 cm | |
53.8 cm | |
42.1 cm |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.