Learn More
Labconco™ WaterPro™ BT Water Purification System Accessory, Filter Pack
For use with WaterPro BT Water Purification System
Manufacturer: Labconco™ 9019200/EMD
Description
- Includes twin cartridges with pretreatment, reverse osmosis, activated carbon and deionization filtration
- Required for operation of WaterPro BT Water Purification Systems
- Snap into place and installation is complete
Specifications
21.5 cm | |
Filter Pack | |
Twin cartridges with pretreatment, reverse osmosis, activated carbon and deionization filtration | |
WaterPro BT Water Purification System |
13 in. | |
33 cm | |
2.2 kg |