Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Water for Irrigation, USP, Sterile Grade, Cytiva Hyclone™
Click to view available options
Quantity:
6 x 500 mL
20 L
4 x 4 L
6 x 1 L
200 L
Packaging:
Bioprocess Bag
HDPE Jug
PET Plastic Bottle
Description
- The pH is 5.0 to 7.0
- Complies with the USP monograph for water for irrigation
- Manufactured under cGMP and ISO 13485 guidelines in a FDA registered facility
- Excellent choice for upstream and downstream media/buffer preparation in biopharmaceutical manufacturing applications
- Widely used in medical device manufacturing/processing applications where a high quality, sterile water is required
Specifications
Color | Clear, Colorless |
Endotoxin | <0.25 EU/mL |
Grade | USP |
Packaging | Bioprocess Bag |
Sterility | Sterile |
Certificate of Analysis | Yes |
Quantity | 200 L |
Safety and Handling
ShelfLife | 36 Months |
Recommended Storage | 2-30°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction