Learn More
Cole-Parmer Cole-Parmer Glass Measuring (Mohr) Pipettes, 10 mL; 12/Pk
Supplier:  Cole-Parmer 2556216
Encompass_Preferred
Reusable Mohr measuring pipettes are calibrated "to deliver" to baseline. A "to deliver" pipette must be held vertically with the tip against the side of the receiving vessel and allowed to drain completely. The stated volume is obtained when draining stops. All pipettes are constructed of borosilicate glass, have permanent amber markings, and are treated to reduce chipping. Accuracy is printed on each pipette. Fast, uniform flow times.
Catalog No. NC0480564
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.