Learn More
Sartorius Vivaspin™ 500 Centrifugal Concentrators

One step procedure for small sample amounts
Supplier: Sartorius VS0101/EMD
Description
Vivaspin centrifugal concentrators are disposable ultrafiltration devices for the concentration of biological samples. Vivaspin 500 centrifugal filter units offer a simple, one step procedure for sample concentration with starting volumes up to 500μl. They can effectively be used in fixed angle rotors accepting 2.2 ml centrifuge tubes. The patented vertical membrane design and thin channel filtration chamber (US 5,647,990), minimizes membrane fouling and provides high speed concentrations. Vivaspin 500 units are available with PES membrane in wide range of molecular weight cut-offs. Concentrate recoveries of > 90% can be reached in less than 30 min. The dead stop pocket prevents concentration to dryness.
High flux PES membrane
High retentate recovery > 90 %
Vertical membrane desingn for ultra fast concentration
Universal rotor compatibility
Safe sample concentration due to dead-stop volume
Convenient concentrate recovery by pipetting
Volume graduation for easy reading/monitoring
Specifications
Vivaspin™ 500 Centrifugal Concentrators | |
10,000 | |
500 μL |
PES | |
25/Pk. | |
50 mm |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.