Learn More
Malvern Panalytical Viscosizer - Taylor Dispersion Analysis - Uncoated Capill
Supplier: Malvern Panalytical VSR2009
Malvern Panalytical Viscosizer TD is an automated biophysical characterization tool utilizing Taylor Dispersion Analysis and providing ultra-low volume, solution-based molecular size and stability measurement capabilities, combined with Poiseuille flow for relative viscosity assessment. Viscosizer TD offers biopharmaceutical researchers an orthogonal technique for conformational stability analysis, which is unrivaled in extending into label-free measurements in highly complex solutions, and encompasses a critical size range from small molecules through to protein-based systems.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.