Learn More
Gibco™ Virus Transport Medium
A ready-to-use solution containing Hanks balanced salt solution, heat inactivated FBS, gentamicin sulfate, and amphotericin B.
Supplier: Gibco™ A48750BA
Description
Kit for transporting samples suspected to contain virus
- It is a ready-to-use solution containing Hanks balanced salt solution, heat inactivated FBS, gentamicin sulfate, and amphotericin B
- Suitable for the transport of samples suspected to contain virus
- Kit complies with the formulation published by the Centers for Disease Control and Prevention (CDC) for the preparation of Viral Transport Medium
Specifications
1 X | |
Virus Transport Medium | |
Sterile-filtered | |
1 L |
Liquid | |
Standard Serum Supplementation | |
No Glucose, No Glutamine, No HEPES, No Phenol Red, No Sodium Pyruvate | |
Dry Ice |
Safety and Handling
ShelfLife : 12 months from date of manufacture