Learn More
Remco™ Vikan™ Stiff UST Short Handle Scrubbing Brush
Remco™ Vikan™ Stiff UST Short Handle Scrubbing Brush is deisgned to be an efficient and effective choice for cleaning tough messes in both wet and dry environments.
Supplier: Remco™ 41791
Description
- Angled bristles ideal for cleaning conveyor belts, production lines, machinery, and food preparation surfaces in high risk areas
- The angled and circular arrangement of the Bristle Security Units ensures efficient cleaning
- Color-coded
Specifications
Scrub Brush | |
Stiff | |
Polypropylene | |
FDA Compliant | |
10 in. | |
Short |
Polyester | |
Yes | |
Non-sterile | |
Pink | |
1 Ea. |