Learn More
Remco™ Vikan™ Soft Baker's Brush
Remco™ Vikan™ Soft Baker's Brush has three rows of soft bristles to efficiently sweep dry, fine powders like flour and sugar.
Supplier: Remco™ 45822
Description
- Suitable for dusting large surface areas like tables and equipment
- Bristles are designed for brushing dry, fine particles like dust, flour, and powdered sugar
- The block is molded to increase worker comfort and includes a hanging hole for easy storage
- Color-coded
Specifications
Cleaning Brush | |
Soft | |
Polypropylene | |
FDA Compliant | |
11 in. | |
Medium |
Polyester | |
Yes | |
Non-sterile | |
Green | |
1 Ea. |