Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Remco™ Vikan™ Soft 11 in. Waterfed Deck Scrub

Remco™ Vikan™ Soft 11 in. Waterfed Deck Scrub can be used in a waterfed system or with a standard Euro-thread handle.

Supplier:  Remco™ 70433

 View more versions of this product

Catalog No. 17-227-780


Add to Cart

Description

Description

  • Scrub has effective water distribution
  • Ideal for use in heavily soiled areas such as meat processing plants
  • Color-coded
Specifications

Specifications

Scrub Brush
Soft
Polypropylene
FDA Compliant
Thread type Euro Hygienic
1 Ea.
Polyester
Yes
Non-sterile
Blue
10.9 in.
Large
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit