Learn More
Remco™ Vikan™ Short Handle Washing Brush
Remco™ Vikan™ Short Handle Washing Brush is a short-handled washing brush with split bristles.
Supplier: Remco™ 41946
Description
- Comfortable grip
- Allows you to work for long periods of time without hand fatigue
- Split bristles are great for washing very sensitive areas, like glass, stainless steel, and acrylic surfaces
- Bristles are designed to retain water and chemicals
- Brush can be used holding both the handle and the brush head for better control
- Color-coded
Specifications
Cleaning Brush | |
Soft/Split | |
Polypropylene | |
FDA Compliant | |
9.75 in. | |
Short |
Polyester | |
Yes | |
Non-sterile | |
Yellow | |
1 Ea. |