Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Remco™ Vikan™ Color-Coded Wall Bracket Replacement Hooks

Remco™ Vikan™ Color-Coded Wall Bracket Replacement Hooks combine with a wall bracket system to allow for total customization of your storage systems.

Supplier:  Remco™ 10163

 View more versions of this product

Catalog No. 17-227-012


Add to Cart

Description

Description

  • Contains 4 hooks and 8 pins
  • For use with wall brackets 1017 and 1018
  • Allows total customization of the wall bracket system to fit any color-coded program
  • Extra hooks can be added to hold more hand tools such as brushes, scrapers, and squeegees
  • Hold more smaller hand tools with these easy-to-install hooks
Specifications

Specifications

Hooks
Polypropylene
FDA Compliant
3 x 0.8 x 2.4 in.
Blue
Customizing wall bracket storage systems
1
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit