Learn More
Remco™ Vikan™ Color-Coded Medium Hand Brush
Remco™ Vikan™ Color-Coded Medium Hand Brush is ideal for small hands due to the unique block shape and size.
Supplier: Remco™ 35872
Description
- For scrubbing tables and cutting boards
- Designed for smaller hands, but can be easily used by those with larger hands thanks to the angled design
- Bristles are ideal for cleaning medium and fine particles
- Can be used in both wet and dry applications
Specifications
Hand Brush | |
Yes | |
Non-sterile | |
Green | |
1 Ea. |
Polyester | |
Polypropylene | |
FDA Compliant | |
6.4 in. | |
Medium |