Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Remco™ Vikan™ 3 in. Tube Brush with 3 ft. Flex Handle

Remco™ Vikan™ 3 in. Tube Brush with 3 ft. Flex Handle allows the tube brush to fit in curved pipes easily so the soft bristles can sweep out small particles.

Supplier:  Remco™ 53835

Catalog No. 17-227-459


Add to Cart

Description

Description

  • Designed to clean hard-to-reach areas
  • Soft bristles are attached to a helically wound stainless steel shaft
  • Strong, FDA-compliant, and chemically resistant
  • Can also be used with water and a cleaning agent for a washing effect
  • Color-coded
Specifications

Specifications

Tube Brush
Medium Stiffness
Stainless Steel
FDA Compliant
36.25 in.
Medium
Polyester
Yes
Non-sterile
White
1 Ea.
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit