Learn More
CEM Corporation VIAL CAPS NON-INVASIVE 100/P
SDP
Supplier:  CEM Corporation 908140
Encompass
Septa, IntelliVent, Snap Ring Top, Set of 100. (100) PTFE-Silicon septa used to seal pressure vials. Will seal vials up to 35 Bar (~515 PSI). Snap-on design to be used with vials for Intellivent (non-invasive) pressure sensor design only.
Catalog No. NC9094195