Learn More
TWD Scientific LLC 2 Position Microtitier SBS Format Rack. Case of 100.
SDP
Supplier:  TWD Scientific LLC 5002002CS
Encompass_Preferred
24-position microtitier footprint rack for 1 dram vials (4ml, 15x45mm) & any vials with 15mm OD. Automation Friendly. SBS Format. 100/Case
Catalog No. 50-117-3422