Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Vernier Advanced Chemistry with FSE
Contains 35 experiments appropriate for AP™ Chemistry or college general chemistry.
Supplier: Vernier CHEMA
Description
- For use with Vernier data-collection technology
- Four student alternative versions included for each experiment: computer, LabQuest, calculator, generic version covering all platforms
- Experiments included for Vernier Temperature Probe, pH Sensor, Conductivity Probe, Drop Counter, Colorimeter, Gas Pressure Sensor, Current Probe, ORP Sensor, and Radiation Monitor
- Includes materials list, step-by-step instructions, data tables, questions
- Teacher Information section with directions for setting up experiments, helpful hints, and sample graphs and data
- Word-processing files of student sections on CD
Specifications
For use with Vernier data-collection technology | |
Lab Book |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction