Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Vernier Power Amplifier Accessory Speaker
For use with Power Amplifier.
Supplier: Vernier PAASPAMP
Description
- Includes speaker and accessories
- Used to study mechanical waves and springs
- Contains lightweight metal disc glued to speaker cone
- Attached drive post to metal plate allows you to drive elastic materials such as strings and springs
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction