Learn More
Vernier KidWind simpleGEN Easy-to-build AC Generator Kit
Allows students to explore the basics of electrical generator design.
$91.00 - $556.00
Specifications
For Use With (Application) | Allows students to explore the basics of electrical generator design, demonstrate Faraday's law, light LEDs and how coils, magnets, and rotation affect electrical generation |
---|
Includes: Wind Turbine Hub, Alligator clip cord pair, Digital multimeter, Sandpaper 1/8 sheet, Magnet holder, Ceramic magnet, 8 in. hex shaft with hub quick connect, 4 in. hex shaft, Simple hand generator housing set, Dowels, Red superbright LED, Bi-polar LED bulb and 28 gauge magnet wire with spool
Description
- To demonstrate Faraday's law, light LEDs
- To perform experiments on how coils, magnets, and rotation affect electrical generation
- Can be converted to simple motor by simple modifications
- Also available in classroom pack which includes materials to build 10 generators and to convert two generators to wind turbines
- Recommended for College, High School, and Middle School Science
Activities Performed:
- Generate electricity
- AC power output and measures output voltage
- Compare different designs (coils, magnets, spinning)
- Understanding clean energy
- Exploring energy transformations, relationship between energy and forces
- Collecting, analyzing and interpreting data with advanced tools
- Designing controlled experiments
Warranty and Services
5-year limited warranty (excludes consumables)
Specifications
Allows students to explore the basics of electrical generator design, demonstrate Faraday's law, light LEDs and how coils, magnets, and rotation affect electrical generation |