Learn More
Vernier High Torque Generator with Wires
Produces lower voltage but higher current for a given rotor speed.
Supplier: Vernier KWHIGEN
Description
- Used with water pump load device with higher current draw
- Includes 2mm driveshaft and is interchangeable with either motor
- 4-foot leads with shrink-wrap reinforced soldering
Warranty and Services
1 year
Specifications
High Torque Generator with Wires | |
With 2mm driveshaft, wires, 4-foot leads with shrink-wrap reinforced soldering |