Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Justrite™ VaporTrap™ HDPE, UN/DOT-Certified Carboy with Filter Kit and Six Ports 1/8 in.

Catalog Number p-9072027
Click to view available options
Capacity (Metric):
13.5 L
20 L

For Use With: Safe transport of hazardous liquids; Closure Material: Polypropylene; Color: White

  • For packaging group II and hazardous chemicals
  • Carbon filter prevents volatile organic compound vapors
  • Two-piece cap design
Product Type UN/DOT Carboy with Filter Kit
Capacity (English) 3.6 gal.
Certifications/Compliance UN/DOT
Closure Material Polypropylene
Closure Size 70 mm
Description Six (6) 1/8 in. Tubing OD Ports
Capacity (Metric) 13.5 L
Dimensions (L x W x H) 287 x 235 x 333 mm
Material HDPE
For Use With (Application) Safe transport of hazardous liquids
Height (English) 13.1 in.
Length (English) 11.3 in.
Shape Rectangular
Width (English) 9.25 in.
Color White
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit