Learn More
Vantage Pro2™ Weather Stations
SDPSCI_ED
First weather stations in their class to use frequency-hopping spread-spectrum radio to transmit and receive data up to 1,000 ft. (300m) line of sight.
Supplier:  Davis Instrument Corporation 6152C
Catalog No. S90550A
Add to Cart
Description
Description
Offer great features such as forecasting, on-screen graphing and much more.
  • Quick-view icons show the forecast at a glance
  • A moving ticker-tape display gives additional details
  • Integrated sensor suite combines rain collector, temperature and humidity sensors, and anemometer in one package
  • Humidity sensors are housed inside a radiation shield for improved accuracy, temperature, and humidity
  • Shield protects against solar radiation and other sources of radiated and reflected heat
Specifications
Specifications
23.6 cm
Integrated asensor suite, console
Forecasting, on-screen graphing
14 in.
Vantage Pro2 Cabled Weather Station
35.5 cm
9.3 in.