Learn More
Cole-Parmer Scienceware 38960-0000 PE Cuvette/Cell Washer with 500 mL Receiver Flask
Supplier: Cole-Parmer 8330000
This washer gently and thoroughly cleans cuvettes without the risk of scratching or chipping. Durable polyethylene housing resists most detergents and solvents. Simply connect washer to your vacuum pump or aspirator, place mouth of cell onto rubber cushion to induce suction, and pour water or cleaning fluid into funnel. The fluid will spray forcibly into the cell and drain into the filter flask - eliminating cross contamination and protecting you from hazardous fluids. Air dry ro auto-dry with continued suction.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.