Learn More
Fischer Technical Company Vacuum Pump Classroom Accessory Kit
Vacuum Pump Classroom Accessory Kit
Supplier: Fischer Technical Company LAVDKIT
Alert:
Prop 65
Description
- This kit comes with everything you need to start teaching High Vacuum in the classroom.
- Demonstrate water boiling at room temperature as well as what happens to a balloon at high vacuum plus much more
- Vacuum Pump Accessory Kit includes -1 Ea. acrylic pump plate, 1Ea. 6 x 11 in. glass bell jar, 5 ft. High Vacuum Tubing, 2 hose clamps, 1 quart High Vacuum Pump oil, 3 Ea. balloons, 1 Ea. petri dish. NOTE: Vacuum Pump not included.
- For use with High Vacuum Pumps, rotary vane style.
- Works well with Fischer Technical LAV series.
Certifications
SciEd
Specifications
1 Ea. acrylic pump plate, 1 Ea. 6 x 11 in. glass bell jar, 5 ft. High Vacuum Tubing, 2 hose clamps, 1 quart High Vacuum Pump oil, 3 Ea. balloons, 1 Ea. petri dish | |
High Vacuum | |
High Vacuum Pumps, Rotary Vane Style |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov