Learn More
Cole-Parmer Supco VG64 Digital vacuum gauge, 0 to 12000 µ (0 to 1,600 Pa)
Supplier: Cole-Parmer 6810100
The multi-line LCD features a bar graph that indicates the direction of the vacuum— movement from left to right indicates increasing pressure and in the opposite direction indicates decreasing pressure. Gauge includes an auxiliary port that allows for quick cleaning and drying of the sensor. Use the built-in hanger for added versatility in mounting. Auto shutoff feature turns gauge off after ten minutes when vacuum reads above 12,000 µ (12 Torr). Operates on three AA batteries (not included).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.